Turbidity standards, Stablcal®
Stablcal® reference suspension I acc. Ph. EUR 6.0, 3 NTU, 100 mL
Supplier: Hach
Jump to:
Product Details & Documents
Stablcal® turbidity standards for all turbidimeters. USEPA accepted for calibration purposes. Stablcal® stabilised formazine turbidity standards are true formazine dilutions developed for use in any turbidimeter.
- Precise, pre-mixed formazine solutions
- Based on formazine primary standards
- Enhanced stability saves time and labour
- Confidence in results
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:100 mL
- Turbidity:3 NTU
- Description:Stablcal® reference suspension I acc. Ph. EUR 6.0