Clamp gaskets, HelixMark®
25,40 mm
Catalog # 113-0318
Supplier: Freudenberg Medical Europe GmbH
Jump to:
Product Details & Documents
Silicone, platinum-cured.
- Manufactured according to GMP in a production facility registered by the FDA for the manufacture of medical products
- Can be sterilised by autoclaving, by ethylene oxide and via gamma radiation within the user's validation cycles and sterilisation levels
- Meet USP Class VI requirements. Material certification and lot traceability are included in each box
These biomedical/pharmaceutical grade clamp gaskets promote crevice-free and protrusion-free processing. Contain no contaminants or leachables.
ISO 13485
Certifications: ISO 13485
Specifications
- Ø int.:25,40 mm
- Seq:03