Orders LinkContactUsLinkComponent
Particle traps for use with PLOT columns
Particle traps for use with PLOT columns
Particle trap for PoraPLOT columns, UltiMetal, includes CP-UltiMetal connector, Int.Ø×L: 0,53 mm × 2,5 m
Catalog # VARICP4018
Supplier:   VARIAN
Particle traps for use with PLOT columns
Catalog # VARICP4018
Supplier:  VARIAN
Particle traps for use with PLOT columns
Particle trap for PoraPLOT columns, UltiMetal, includes CP-UltiMetal connector, Int.Ø×L: 0,53 mm × 2,5 m
Price
253,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Though highly stabilised, it is impossible to guarantee that no particles will dislodge from the column wall. When used in valve-switching applications, the use of a particle trap can prevent scarring of the rotors.
Specifications
  • Description:
    Particle trap for PoraPLOT columns, UltiMetal, includes CP-UltiMetal connector, Int.Ø×L: 0,53 mm × 2,5 m
  • Seq:
    05