Order Entry
Germany
Orders LinkContactUsLinkComponent
Analytical HPLC columns, Microsorb™ Si

Analytical HPLC columns, Microsorb™ Si

Analytical, 100 Å, 3 µm, 4,60 mm, 100 mm
Catalog # VARIR0089100E3
Supplier:   VARIAN
Analytical HPLC columns, Microsorb™ Si
Catalog # VARIR0089100E3
Supplier:  VARIAN
Analytical HPLC columns, Microsorb™ Si

Analytical, 100 Å, 3 µm, 4,60 mm, 100 mm

Price
882,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Microsorb™ Silica is ideal for the separation of low molecular weight compounds that dissolve in organic solvents. Separations on silica columns depend upon the difference in orientation, type and number of functional groups associated with the compounds in the sample.

Specifications
  • Column Format:
    Analytical
  • Ø int.:
    4,60 mm
  • Particle Size:
    3 µm
  • Pore size:
    100 Å
  • Length:
    100 mm
  • Seq:
    01