Analytical HPLC columns, Microsorb™ Si
Analytical, 100 Å, 5 µm, 4,60 mm, 150 mm
Catalog # VARIR0089100D5
Supplier: VARIAN
Jump to:
Product Details & Documents
Microsorb™ Silica is ideal for the separation of low molecular weight compounds that dissolve in organic solvents. Separations on silica columns depend upon the difference in orientation, type and number of functional groups associated with the compounds in the sample.
Specifications
- Column Format:Analytical
- Ø int.:4,60 mm
- Particle Size:5 µm
- Pore size:100 Å
- Length:150 mm
- Seq:02



