Analytical HPLC columns, Microsorb™ Si
Analytical, 100 Å, 5 µm, 4,60 mm, 250 mm
Catalog # VARIR0089100C5
Supplier: VARIAN
Jump to:
Product Details & Documents
Microsorb™ Silica is ideal for the separation of low molecular weight compounds that dissolve in organic solvents. Separations on silica columns depend upon the difference in orientation, type and number of functional groups associated with the compounds in the sample.
Specifications
- Column Format:Analytical
- Ø int.:4,60 mm
- Particle Size:5 µm
- Pore size:100 Å
- Length:250 mm
- Seq:03
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



