Solid phase extraction cartridges, Bond Elut™ Florisil
Bond Elut™ Jr, Florisil, 200 µm, 1000 mg
Catalog # VARI12166014B
Supplier: VARIAN
Jump to:
Product Details & Documents
Florisil is a magnesia-loaded silica gel, extremely polar in nature and ideal for the isolation of polar compounds from non-polar matrices.
- Pesticide residue (PR) grade
- Fast flow, ideal for viscous samples
The larger particle size of the sorbent enables flow for large sample volumes and is therefore an attractive alternative to silica if the sample matrix is particularly viscous.
Specifications
- Particle Size:200 µm
- Description:Bond Elut™ Jr
- Seq:15
- Bed weight:1000 mg
- Phase:Florisil