Orders LinkContactUsLinkComponent
Particle traps for use with PLOT columns
Particle traps for use with PLOT columns
Particle trap connectors for PoraPLOT columns, fused silica, 0,25/0,32 mm ID
Catalog # VARICP4788
Supplier:   VARIAN
Particle traps for use with PLOT columns
Catalog # VARICP4788
Supplier:  VARIAN
Particle traps for use with PLOT columns
Particle trap connectors for PoraPLOT columns, fused silica, 0,25/0,32 mm ID
Price
224,00 €
Log in for availability
1 * 10 ST
Product Details & Documents
Though highly stabilised, it is impossible to guarantee that no particles will dislodge from the column wall. When used in valve-switching applications, the use of a particle trap can prevent scarring of the rotors.
Specifications
  • Description:
    Particle trap connectors for PoraPLOT columns, fused silica, 0,25/0,32 mm ID
  • Seq:
    06