Cell counter vials
Cell counter vial 1 with lid, for Coulter® and Hycel® cell counter systems, 25,00 ml, PS, Clear, Yes, 250 vials/bag, 4 bags
Catalog # RATI5515040
Supplier: RATIOLAB
Jump to:
Product Details & Documents
Cell counter vials made of clear PS.
- High-precision design for use with robotic analytical systems
Specifications
- Cap included:Yes
- Packed:250 vials/bag, 4 bags
- Capacity:25,00 ml
- Colour:Clear
- Description:Cell counter vial 1 with lid, for Coulter® and Hycel® cell counter systems
- Material:PS
- Seq:02