pH buffer solutions, referenced at 20 °C, Fluka™
12,00, ±0.05, Phosphate, 20 °C, Plastic bottle, 1×100 ml, 100 mL
Supplier: Honeywell Chemicals
pH buffer solutions, referenced at 20 °C, Fluka™
Catalog # 33651-100ML
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
Documents
Specifications
Specifications
- Pack type:Plastic bottle
- pH buffer type:Phosphate
- Pk:100 mL
- Reference temperature:20 °C
- pH accuracy:±0.05
- Pk Info:1×100 ml
- pH value:12,00



