Order Entry
Germany
Orders LinkContactUsLinkComponent
pH buffer solutions, referenced at 20 °C, Fluka™

pH buffer solutions, referenced at 20 °C, Fluka™

11,00, ±0.05, Borate, 20 °C, Plastic bottle, 1×500 ml, 500 mL
Catalog # 33650-500ML
Supplier:   Honeywell Chemicals
pH buffer solutions, referenced at 20 °C, Fluka™
Catalog # 33650-500ML
CAS Number:  
pH buffer solutions, referenced at 20 °C, Fluka™

11,00, ±0.05, Borate, 20 °C, Plastic bottle, 1×500 ml, 500 mL

CAS Number:  
Price
54,60 €
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Borate
  • Pk:
    500 mL
  • Reference temperature:
    20 °C
  • pH accuracy:
    ±0.05
  • Pk Info:
    1×500 ml
  • pH value:
    11,00

Specifications