Order Entry
Germany
Orders LinkContactUsLinkComponent
Hydroxyzine Related Compounds, Reference Standard

Hydroxyzine Related Compounds, Reference Standard

Hydroxyzine Hydrochloride, USP reference standard, Vial, 500 mg
Catalog # 1333003.
Supplier:   USP
Hydroxyzine Related Compounds, Reference Standard
Catalog # 1333003.
Supplier:  USP
CAS Number:  
Hydroxyzine Related Compounds, Reference Standard

Hydroxyzine Hydrochloride, USP reference standard, Vial, 500 mg

CAS Number:  
Price
404,00 €
Log in for availability
1 * 500 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Documents

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    500 mg
  • Standards:
    USP reference standard
  • Description:
    Hydroxyzine Hydrochloride
  • Cat. No.:
    1333003.
  • Formula:
    C₂₁H₂₉Cl₃N₂O₂
  • Description:
    Hydroxyzine Hydrochloride
  • CAS No.:
    2192-20-3

Specifications