Order Entry
Germany
Orders LinkContactUsLinkComponent
Halogen lamps

Halogen lamps

Halogen fibre optic lamp, with smooth reflector finish, Dental, microscope and medical applications, Osram 64255, G4, 8 V, 20 W
Catalog # ACEA64255
Supplier:   Ace Audio Visual
Halogen lamps
Catalog # ACEA64255
Supplier:  Ace Audio Visual
Halogen lamps
Halogen lamps

Halogen fibre optic lamp, with smooth reflector finish, Dental, microscope and medical applications, Osram 64255, G4, 8 V, 20 W

Price
44,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • For:
    Dental, microscope and medical applications
  • Description:
    Halogen fibre optic lamp, with smooth reflector finish
  • Seq:
    08
  • Base:
    G4
  • Model:
    Osram 64255
  • Power:
    20 W
  • Voltage:
    8 V
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.