Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
FILTER FLUORO 584NM EXCIT HBW 16NM 1 * 1 items
Catalog # LABL1425842
Supplier:
THERMO LABSYSTEMS LIFE SCIENCE
Expand image
FILTER FLUORO 584NM EXCIT HBW 16NM 1 * 1 items
Catalog # LABL1425842
Supplier:
THERMO LABSYSTEMS LIFE SCIENCE