Order Entry
Germany
Orders LinkContactUsLinkComponent
Reaction vessels with frame assemblies

Reaction vessels with frame assemblies

6000 ml, flange NW 150, 425 mm, DN 15
Catalog # KGWARK-FB-17
Supplier:   KGW
Reaction vessels with frame assemblies
Catalog # KGWARK-FB-17
Supplier:  KGW
Reaction vessels with frame assemblies

6000 ml, flange NW 150, 425 mm, DN 15

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Reaction vessel with stationary frame, J valve and accessories

Specifications
  • Connection:
    DN 15
  • With:
    flange NW 150
  • Capacity:
    6000 ml
  • Height:
    425 mm
  • Seq:
    02