Estriol
Glass bottle for solids, 100 mg
Catalog # CAYM10006484-100
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
- CAS Number:50-27-1
- Formula:C₁₈H₂₄O₃
- MDL Number:MFCD00003691
- EINECS:200-022-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



