Estradiol
Glass bottle for solids, 500 mg
Catalog # CAYM10006315-500
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: beta-Estradiol, 17β-Estradiol
- CAS Number:50-28-2
- Formula:C₁₈H₂₄O₂
- MDL Number:MFCD00003693
- Molecular Weight:272.39 g/mol
- EINECS:200-023-8
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg



