Order Entry
Germany
Orders LinkContactUsLinkComponent
Tissue processing/embedding cassettes, Histosette® II, M492

Tissue processing/embedding cassettes, Histosette® II, M492

Orange
Catalog # 720-0907
Supplier:   Simport Scientific
Tissue processing/embedding cassettes, Histosette® II, M492
Catalog # 720-0907
Supplier:  Simport Scientific
Tissue processing/embedding cassettes, Histosette® II, M492
Tissue processing/embedding cassettes, Histosette® II, M492

Orange

Price
336,00 €
Log in for availability
1 * 1.500 ST
Product Details & Documents

Acetal polymer.

  • Resistant to the chemical action of histological solvents
  • Flow-through slots maximise fluid exchange
  • Integral lid
  • Anterior writing area at 45° angle to enable use with leading cassette labelling instruments
Specifications
  • Colour:
    Orange
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.