MINICYL regulators
MINICYL regulator, 6,3 mm brass fittings, max pressure: 300 psi
Catalog # REST20611
Supplier: Restek
Jump to:
Product Details & Documents
This compact general purpose regulator is constructed of lightweight aluminum with an elastomer diaphragm. It has many laboratory applications including air-drying glassware, sparging or evaporating solutions and controlling pneumatic valves.
Specifications
- Description:MINICYL regulator, 6,3 mm brass fittings, max pressure: 300 psi
- Seq:02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



