Orders LinkContactUsLinkComponent
Nuts and ferrules
Nuts and ferrules
Ferrule, 1/16", Stainless steel
Catalog # REST20286
Supplier:   Restek
Nuts and ferrules
Catalog # REST20286
Supplier:  Restek
Nuts and ferrules
Nuts and ferrules
Ferrule, 1/16", Stainless steel
Price
94,44 €
Log in for availability
1 * 10 ST
Product Details & Documents
These nuts and ferrules are used for Valco® connectors.
Specifications
  • Description:
    Ferrule, 1/16"
  • Material:
    Stainless steel
  • Seq:
    01