Flow controllers, Veriflo
Stainless steel, Flow controller, without orifice
Catalog # REST24239
Supplier: Restek
Jump to:
Product Details & Documents
Veriflo controllers can be used with passive air samplers in maintaining a constant flow rate.
Entire sample flow path is made of stainless steel or siltek® treated for maximum inertness.
Specifications
- Description:Flow controller, without orifice
- Material:Stainless steel
- Seq:01