Flow controllers, Veriflo
Siltek® behandeltes Edelstahl, Flow controller, without orifice
Catalog # REST24238
Supplier: Restek
Jump to:
Product Details & Documents
Veriflo controllers can be used with passive air samplers in maintaining a constant flow rate.
Entire sample flow path is made of stainless steel or siltek® treated for maximum inertness.
Specifications
- Bezeichnung:Flow controller, without orifice
- Material:Siltek® behandeltes Edelstahl
- Seq:02