Orders LinkContactUsLinkComponent
Caps for screw thread vials
Open-hole caps, PP, black
Catalog # REST21177
Supplier:   Restek
Caps for screw thread vials
Catalog # REST21177
Supplier:  Restek
Caps for screw thread vials
Caps for screw thread vials
Open-hole caps, PP, black
Price
92,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
Black PP, open hole caps fit all 2,0 ml, 12×32 mm, screw-thread 8 mm/425 vial-based autosamplers.
Specifications
  • Description:
    Open-hole caps, PP, black
  • Seq:
    02