D-Cycloserine
Glass bottle for solids, 5 g
Catalog # MOLE69907662-5G
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: D-(+)-Cycloserine, Cycloserine, D(+)-Cycloserine, (R)-4-Aminoisoxazolidin-3-one, D-4-Aminoisoxazolidin-3-one, (R)-(+)-Cycloserine
- CAS Number:68-41-7
- Formula:C₃H₆N₂O₂
- MDL Number:MFCD00005353
- Molecular Weight:102.09 g/mol
- Storage Temperature:Freezer
- EINECS:200-688-4
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



