Order Entry
Germany
Orders LinkContactUsLinkComponent
Ammonia solution 9,9 - 10,3% analytical reagent

Ammonia solution 9,9 - 10,3% analytical reagent

Glass bottle, 2.500 mL
Catalog # KRAF06212.4010
Corrosive
Irritant
Environment
Supplier:   Bernd Kraft
Ammonia solution 9,9 - 10,3% analytical reagent
Catalog # KRAF06212.4010
Supplier:  Bernd Kraft
CAS Number:  1336-21-6
Ammonia solution 9,9 - 10,3% analytical reagent

Glass bottle, 2.500 mL

CAS Number:  1336-21-6
Price
69,00 €
Log in for availability
1 * 2.500 mL
Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Density:0.92 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2.500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.