4-Chloro-3-fluorophenol
Glass bottle for solids, 10 g
Catalog # FLUO004459-10G
Marketsource
Supplier: FLUOROCHEM
4-Chloro-3-fluorophenol
Catalog # FLUO004459-10G
Supplier: FLUOROCHEM
Marketsource
CAS Number: 348-60-7
Jump to:
Product Details & Documents
- CAS Number:348-60-7
- Formula:C₆H₄ClFO
- MDL Number:MFCD00042583
- Molecular Weight:146.55 g/mol
- Boiling Pt:163 °C (1013 hPa)
- Melting Pt:49…53 °C
- UN:3261
- ADR:8,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



