Order Entry
Germany
Orders LinkContactUsLinkComponent
2,4,6-Trifluorophenol

2,4,6-Trifluorophenol

Glass bottle for solids, 1 g
Catalog # FLUO001898-1G
Marketsource
Supplier:   FLUOROCHEM
2,4,6-Trifluorophenol
Catalog # FLUO001898-1G
Supplier:  FLUOROCHEM
Marketsource
CAS Number:  2268-17-9
2,4,6-Trifluorophenol

Glass bottle for solids, 1 g

CAS Number:  2268-17-9
Price
51,10 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:2268-17-9
  • Formula:C₆H₃F₃O
  • MDL Number:MFCD00061216
  • Molecular Weight:148.08 g/mol
  • Boiling Pt:155 °C (1013 hPa)
  • Melting Pt:50 °C
  • UN:3261
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications