Order Entry
Germany
Orders LinkContactUsLinkComponent
ω,4-Dibromoacetophenone

ω,4-Dibromoacetophenone

Glass bottle for solids, 25 g
Catalog # FLUO001372-25G
Corrosive
Marketsource
Supplier:   FLUOROCHEM
ω,4-Dibromoacetophenone
Catalog # FLUO001372-25G
Supplier:  FLUOROCHEM
Marketsource
CAS Number:  99-73-0
ω,4-Dibromoacetophenone

Glass bottle for solids, 25 g

CAS Number:  99-73-0
Price
99,20 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Tau Protein Kinase I Inhibitor, 2,4'-Dibromoacetophenone, Glycogen Synthase Kinase-3β Inhibitor VII, α,4-Dibromoacetophenone, GSK-3β Inhibitor VII, TPK I Inhibitor, 2-Bromo-1-(4-bromophenyl)-1-ethanone, 4-Bromophenacyl bromide
  • CAS Number:99-73-0
  • Formula:C₈H₆Br₂O
  • MDL Number:MFCD00000200
  • Molecular Weight:277.94 g/mol
  • Merck Index:13,01412
  • Boiling Pt:310 °C (1013 hPa)
  • Melting Pt:107…108 °C
  • Storage Temperature:Refrigerator
  • EINECS:202-783-6
  • UN:3261
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.