ω,4-Dibromoacetophenone
Glass bottle for solids, 100 g
Catalog # FLUO001372-100G
Marketsource
Supplier: FLUOROCHEM
ω,4-Dibromoacetophenone
Catalog # FLUO001372-100G
Supplier: FLUOROCHEM
Marketsource
CAS Number: 99-73-0
Jump to:
Product Details & Documents
Synonyms: Tau Protein Kinase I Inhibitor, 2,4'-Dibromoacetophenone, Glycogen Synthase Kinase-3β Inhibitor VII, α,4-Dibromoacetophenone, GSK-3β Inhibitor VII, TPK I Inhibitor, 2-Bromo-1-(4-bromophenyl)-1-ethanone, 4-Bromophenacyl bromide
- CAS Number:99-73-0
- Formula:C₈H₆Br₂O
- MDL Number:MFCD00000200
- Molecular Weight:277.94 g/mol
- Merck Index:13,01412
- Boiling Pt:310 °C (1013 hPa)
- Melting Pt:107…108 °C
- Storage Temperature:Refrigerator
- EINECS:202-783-6
- UN:3261
- ADR:8,II
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Cat. No.:FLUO001372-100G



