Order Entry
Germany
Orders LinkContactUsLinkComponent
4-Fluorophenol

4-Fluorophenol

Glass bottle for solids, 1 kg
Catalog # FLUO001291-1KG
Irritant
Marketsource
Supplier:   FLUOROCHEM
4-Fluorophenol
Catalog # FLUO001291-1KG
Supplier:  FLUOROCHEM
Marketsource
CAS Number:  371-41-5
4-Fluorophenol

Glass bottle for solids, 1 kg

CAS Number:  371-41-5
Price
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  p-Fluorophenol, 1-Fluoro-4-hydroxybenzene
  • CAS Number:371-41-5
  • Formula:C₆H₅FO
  • MDL Number:MFCD00002316
  • Molecular Weight:112.1 g/mol
  • Boiling Pt:185 °C (1013 hPa)
  • Melting Pt:44…47 °C
  • Density:1.22 g/cm³ (20 °C)
  • Flash Pt:68 °C
  • Storage Temperature:Ambient
  • EINECS:206-736-0
  • UN:2923
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications