(2-Bromophenyl)acetonitrile
Glass bottle for solids, 100 g
Catalog # FLUO001135-100G
Marketsource
Supplier: FLUOROCHEM
(2-Bromophenyl)acetonitrile
Catalog # FLUO001135-100G
Supplier: FLUOROCHEM
Marketsource
CAS Number: 19472-74-3
Jump to:
Product Details & Documents
Synonyms: 2-Bromobenzyl cyanide
- CAS Number:19472-74-3
- Formula:C₈H₆BrN
- MDL Number:MFCD00001896
- Molecular Weight:196.05 g/mol
- Boiling Pt:270 °C (1013 hPa)
- Melting Pt:1 °C
- Density:1.51 g/cm³ (20 °C)
- EINECS:243-091-4
- UN:1694
- ADR:6.1,I
- :Density: 1.51 g/cm³ (20 °C)
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



