Order Entry
Germany
Orders LinkContactUsLinkComponent
2-Hydroxy-4-nitrobenzonitrile

2-Hydroxy-4-nitrobenzonitrile

Glass bottle for solids, 5 g
Catalog # FLUO012255-5G
Marketsource
Supplier:   FLUOROCHEM
2-Hydroxy-4-nitrobenzonitrile
Catalog # FLUO012255-5G
Supplier:  FLUOROCHEM
Marketsource
CAS Number:  39835-14-8
2-Hydroxy-4-nitrobenzonitrile

Glass bottle for solids, 5 g

CAS Number:  39835-14-8
Price
223,00 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:39835-14-8
  • Formula:C₇H₄N₂O₃
  • MDL Number:MFCD02258875
  • Molecular Weight:164.1183 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.