3'-Nitroformanilide
Glass bottle for solids, 25 g
Catalog # FLUO011202-25G
Marketsource
Supplier: FLUOROCHEM
Jump to:
Product Details & Documents
Synonyms: 3-Nitroformanilide
- CAS Number:102-38-5
- Formula:C₇H₆N₂O₃
- MDL Number:MFCD00017014
- Molecular Weight:166.14 g/mol
- Melting Pt:134 °C
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



