Order Entry
Germany
Orders LinkContactUsLinkComponent
 

3-(2-Fluorophenyl)propionic acid

Glass bottle for solids, 1 g
Catalog # FLUO009235-1G
Marketsource
Supplier:   FLUOROCHEM
3-(2-Fluorophenyl)propionic acid
Catalog # FLUO009235-1G
Supplier:  FLUOROCHEM
Marketsource
CAS Number:  1643-26-1
3-(2-Fluorophenyl)propionic acid

Glass bottle for solids, 1 g

CAS Number:  1643-26-1
Price
32,10 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:1643-26-1
  • Formula:C₉H₉FO₂
  • MDL Number:MFCD01310820
  • Molecular Weight:168.17 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications