Order Entry
Germany
Orders LinkContactUsLinkComponent
 

4'-(4-Fluorophenoxy)acetophenone

Glass bottle for solids, 5 g
Catalog # FLUO023222-5G
Marketsource
Supplier:   FLUOROCHEM
4'-(4-Fluorophenoxy)acetophenone
Catalog # FLUO023222-5G
Supplier:  FLUOROCHEM
Marketsource
CAS Number:  35114-93-3
4'-(4-Fluorophenoxy)acetophenone

Glass bottle for solids, 5 g

CAS Number:  35114-93-3
Price
268,00 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:35114-93-3
  • Formula:C₁₄H₁₁FO₂
  • MDL Number:MFCD02246922
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications