Order Entry
Germany
Orders LinkContactUsLinkComponent
Cavity slides

Cavity slides

Cavity slides, Clear, 1, 3 - 4 mm, 22 mm
Catalog # HECH42415010
Cavity slides
Catalog # HECH42415010
Cavity slides
Cavity slides

Cavity slides, Clear, 1, 3 - 4 mm, 22 mm

Price
159,00 €
Log in for availability
1 * 10 ST
Product Details & Documents

Cavity microscopy slides.

  • Ground edges
  • Made from clear glass

Suitable for staining applications in microscopy.

Specifications
  • No. of wells:
    1
  • Colour:
    Clear
  • Description:
    Cavity slides
  • Seq:
    04
  • Thickness:
    3 - 4 mm
  • Well Ø:
    22 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.