Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Quick-dry foam squeegee

Quick-dry foam squeegee, 24”, 609,6 mm
Catalog # 129-0098
Supplier:   PERFEX
Quick-dry foam squeegee
Catalog # 129-0098
Supplier:  PERFEX
Quick-dry foam squeegee
Quick-dry foam squeegee

Quick-dry foam squeegee, 24”, 609,6 mm

Price
29,20 €
Log in for availability
1 * 1 ST
Product Details & Documents

These twin-blade neoprene rubber squeegees are specifically designed for use on uneven and irregular surfaces. Features a durable polymer channel and flexible blades that quickly wipe surfaces dry. Lightweight, non-marring, and easy to clean with warm water.

  • Light weight and non-marring
  • Easy to clean with warm water
  • Easy blade change system

Resistant to water, oil, grease, detergents, sanitisers, and solvents.

Note: Not recommended for abrasive surfaces.

 : Polymer adjustable or fixed handle must be ordered separately.

Specifications
  • Description:
    Quick-dry foam squeegee, 24”
  • Length:
    609,6 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.