Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
pH-METER ECOSCAN PH6 PH/MV+TPROBE 1 * 1 items
Catalog # EUTEEC-PH6/02K
Supplier:
EUTECH
Expand image
pH-METER ECOSCAN PH6 PH/MV+TPROBE 1 * 1 items
Catalog # EUTEEC-PH6/02K
Supplier:
EUTECH