Orders LinkContactUsLinkComponent
Stirrer guides
Stirrer guides
Stirrer guide, with PP-insert, 29/32, — mm
Catalog # WITG1.470.029
Supplier:   witeg Labortechnik
Stirrer guides
Catalog # WITG1.470.029
Supplier:  witeg Labortechnik
Stirrer guides
Stirrer guides
Stirrer guide, with PP-insert, 29/32, — mm
Price
80,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stirrer guides are of different types.
With fused-on stirrer bearing.
GTS-vacuum-stirrer, with PTFE-insert and interchangeable PTFE-vacuum-seal, screw cap GL 25.
With PP-insert, 8 mm Ø and screw cap GL 25, made of melamine.
With cooling jacket and PTFE-insert, with interchangeable PTFE-vacuum seal and screw cap GL 25, made of melamine.
Specifications
  • NS:
    29/32
  • Description:
    Stirrer guide, with PP-insert
  • Seq:
    04
  • Shaft Ø:
    — mm