Fractionating columns, Vigreux
300 mm, 24/29 NS, 24/29 NS
Catalog # 201-0177
Supplier: witeg Labortechnik
Jump to:
Product Details & Documents
Borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With or without glass jacket.
Specifications
- Cone size:24/29 NS
- Socket size:24/29 NS
- Total length:300 mm