Stirrer guides
Stirrer guide, with fused-on stirrer bearing, 29/32, 10 mm
WITG1.460.029
Stirrer guides are of different types.
With fused-on stirrer bearing.
GTS-vacuum-stirrer, with PTFE-insert and interchangeable PTFE-vacuum-seal, screw cap GL 25.
With PP-insert, 8 mm Ø and screw cap GL 25, made of melamine.
With cooling jacket and PTFE-insert, with interchangeable PTFE-vacuum seal and screw cap GL 25, made of melamine.
- NS:29/32
- Description:Stirrer guide, with fused-on stirrer bearing
- Seq:01
- Shaft Ø:10 mm



