Glass beads
Glass beads, solid, 6 mm
Catalog # 201-0280
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Solid glass.
For packing distillation columns.
Specifications
- Ø:6 mm
- Description:Glass beads, solid
- Seq:07