Glass beads
Glass beads, solid, 4 mm
Catalog # 201-0278
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Solid glass.
For packing distillation columns.
Specifications
- Ø:4 mm
- Description:Glass beads, solid
- Seq:04