Bellow connectors
45/40 NS, 170 mm, 45/40 NS
BOHLH906-14
Bellow connectors, made of PTFE, with sharp folds and sealing rings on the outside of the cone.
- Universal chemical resistance
- Suitable for vacuum applications
- Temperature resistance from −200 to +250 °C
- Compensates vibrations from vacuum pumps
- Suiltable for length compensation of heated columns and angular misalignment
- Cone size:45/40 NS
- Socket size:45/40 NS
- Seq:05
- Total length:170 mm



