Orders LinkContactUsLinkComponent
Pipette tips
Extended, 50 - 1200 µl, +, 90 mm, Purple, 10 racks, 96 each
  613-5111
   Low Manufacturing Impact
 :   Sartorius
Pipette tips
  613-5111
 :  Sartorius
   Low Manufacturing Impact
Pipette tips
Pipette tips
Extended, 50 - 1200 µl, +, 90 mm, Purple, 10 racks, 96 each
133,00 €
1 * 960 ST
These pipette tips are suitable for many pipettes on the market.
  • Natural colour, premium grade PP, autoclavable
  • Excellent seal design for accurate, leak-tight sealing
  • Produced and packed in a dust-free environment under strictly maintained conditions
 : Available in bulk packaging, in autoclavable racks and in refills.
 : Article 613-4153 (BHIT780310) is designed for use with the Midi Plus Pipette Controller only.
  • Colour code:
    Purple
  • Packed:
    10 racks, 96 each
  • Sterile:
    +
  • Volume:
    50 - 1200 µl
  • Length:
    90 mm
  • Seq:
    34
  • Tip style:
    Extended
  • Cat. No.:
    613-5111
  • Environmental certification:
    Renewable energy certificate; ISO 14001
  • Environmentally Preferable:
    Low Manufacturing Impact