Order Entry
Germany
Orders LinkContactUsLinkComponent
Ammonia solution 25%, extra pure

Ammonia solution 25%, extra pure

Glass bottle, 1 L
Catalog # KRAF06010.3010
Corrosive
Irritant
Environment
Supplier:   Bernd Kraft
Ammonia solution 25%, extra pure
Catalog # KRAF06010.3010
Supplier:  Bernd Kraft
CAS Number:  1336-21-6
Ammonia solution 25%, extra pure

Glass bottle, 1 L

CAS Number:  1336-21-6
Price
24,50 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Boiling Pt:32…37.7 °C (1013 hPa)
  • Melting Pt:–60…–57.5 °C
  • Density:0.88…0.91 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.