Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
EV243 Electrophoresis power supply/ pack 400V, 300 mA, 50W 1 * 1 items
Catalog # 730-1096
Supplier:
CONSORT
Expand image
EV243 Electrophoresis power supply/ pack 400V, 300 mA, 50W 1 * 1 items
Catalog # 730-1096
Supplier:
CONSORT