Order Entry
Germany
Orders LinkContactUsLinkComponent
Test tubes with screw cap

Test tubes with screw cap

12 ml, 102 mm, 16 mm, Sterile
Catalog # KART0560100
Supplier:   KARTELL
Test tubes with screw cap
Catalog # KART0560100
Supplier:  KARTELL
Test tubes with screw cap
Test tubes with screw cap

12 ml, 102 mm, 16 mm, Sterile

Price
1 450,00 €
Log in for availability
1 * 1.500 ST
Product Details & Documents

PS, clear, with PE screw cap.

  • Good thermal resistance
  • Good chemical resistance

Tubes have a round bottom and a freestanding skirt. Sterile tubes are available individually wrapped. The 30 ml individual tubes have a self-adhesive label for marking.

Specifications
  • Ext.Ø:
    16 mm
  • Sterility:
    Sterile
  • Capacity:
    12 ml
  • Length:
    102 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.