Orders LinkContactUsLinkComponent
Pycnometers, unadjusted
Pycnometers, unadjusted
2 ml
Catalog # WITG3.900.002
Supplier:   witeg Labortechnik
Pycnometers, unadjusted
Catalog # WITG3.900.002
Supplier:  witeg Labortechnik
Pycnometers, unadjusted
Pycnometers, unadjusted
2 ml
Price
24,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.
  • With ground-in capillary stopper
  • Nominal capacity printed on the bottom
  • Very good chemical and temperature resistance
Certifications: DIN 12797
Specifications
  • Volume (nom.):
    2 ml