ContactUsLinkComponent
TNFalpha, Rat, Biotrak Assay, ELISA - 96 wells (Range: 31-2500pg/ml. Sensitivity: <15pg/ml. Sample volume: 50 ul. 1.5L wash buffer s upplied. Suitable for use with serum, plasma, and cell culture supernatant samples) 1 * 1 KIT
Catalog # RPN2744
Supplier:  Cytiva
TNFalpha, Rat, Biotrak Assay, ELISA - 96 wells (Range: 31-2500pg/ml. Sensitivity: <15pg/ml. Sample volume: 50 ul. 1.5L wash buffer s upplied. Suitable for use with serum, plasma, and cell culture supernatant samples) 1 * 1 KIT
Catalog # RPN2744
Supplier:  Cytiva
Frequently Bought Together